Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4953MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: ODC1/485. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: PC3 cells. Placenta, Prostate or Testicular carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human ODC-1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 53kDa.Chromosome: 2p25.1. Gene Symbol: ODC1. Gene ID: 4953. Ensemble: ENSG00000115758. Uniprot: NP_002530.
Catalog No. 50-188-5026