Learn More
Medchemexpress LLC HY-109120 5mg , Odevixibat CAS:501692-44-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1091205MG
Encompass_Preferred
HY-109120 5mg Odevixibat CAS: 501692-44-0 Odevixibat (A4250) is a potent, selective and orally active inhibitor of the ileal bile acid transporter (IBAT). Odevixibat (A4250)decreases cholestatic liver and bile duct injury in mice model. Odevixibat (A4250) has the potential for the treatment of primary biliary cirrhosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9928