Learn More
Aviva Systems Biology Corp Odf1 Antibody - middle region (ARP53765_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53765_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Odf1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-880