Learn More
Aviva Systems Biology Corp ODF2 Antibody - N-terminal region (ARP53831_P050) | 100uL
SDP
Catalog No. 5001462967
Encompass_Preferred
This is a rabbit polyclonal antibody against ODF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ODF2. Alternate names: FLJ44866, MGC111096, MGC9034, ODF2/1, ODF2/2, ODF84, CT134ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62967 Supplier Aviva Systems Biology Corp Supplier No. ARP53831_P050
This is a rabbit polyclonal antibody against ODF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ODF2. Alternate names: FLJ44866, MGC111096, MGC9034, ODF2/1, ODF2/2, ODF84, CT134ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat