Learn More
Aviva Systems Biology Corp ODF3L1 Antibody - N-terminal region (ARP53333_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53333_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ODF3L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ODF3L1. Alternate names: MGC48986ApplicationWB. Species reactivity: Guinea Pig, Human, Rat
Catalog No. 50-014-61000