Learn More
Aviva Systems Biology Corp ODF4 Antibody - N-terminal region (ARP53127_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53127_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ODF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ODF4. Alternate names: MGC138215, MGC138219, OPPO1, CT134, CT136ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60936