Learn More
MSC ODIN FOG-OFF SFTY GLS
Catalog No. 5000677207
Encompass
These safety glasses feature anti-fog and scratch-resistant clear polycarbonate lenses, designed with a full-framed, wrap-around style for universal sizing. They provide UV protection and impact resistance, meeting ANSI Z87.1+, CSA Z94.3, and MIL-PRF 32432 Ballistic Impact standards. The gray frame is made of rubber with non-adjustable temples, and anti-fog coatings enhance visibility in hot and humid conditions.
  • Anti-fog and scratch-resistant lens coating
  • Full-framed, wrap-around design
  • Clear polycarbonate lenses
  • Gray rubber frame and temples
  • Universal size with non-adjustable temples
  • UV protection
  • Impact-resistant
  • Meets ANSI Z87.1+, CSA Z94.3, and MIL-PRF 32432 ballistic impact standards
Catalog No. 50-006-77207 Supplier MSC Supplier No. 31782295
These safety glasses feature anti-fog and scratch-resistant clear polycarbonate lenses, designed with a full-framed, wrap-around style for universal sizing. They provide UV protection and impact resistance, meeting ANSI Z87.1+, CSA Z94.3, and MIL-PRF 32432 Ballistic Impact standards. The gray frame is made of rubber with non-adjustable temples, and anti-fog coatings enhance visibility in hot and humid conditions.
  • Anti-fog and scratch-resistant lens coating
  • Full-framed, wrap-around design
  • Clear polycarbonate lenses
  • Gray rubber frame and temples
  • Universal size with non-adjustable temples
  • UV protection
  • Impact-resistant
  • Meets ANSI Z87.1+, CSA Z94.3, and MIL-PRF 32432 ballistic impact standards