Learn More
TARGETMOL CHEMICALS INC ODM-203 25MG
SDP
Catalog No. 502579174
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ODM-203 a Selective Inhibitor of FGFR and VEGFR Shows Strong Antitumor Activity and Induces Antitumor Immunitypurity: 98%
Catalog No. 50-257-9174 Supplier TARGETMOL CHEMICALS INC Supplier No. T761125MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ODM-203 a Selective Inhibitor of FGFR and VEGFR Shows Strong Antitumor Activity and Induces Antitumor Immunitypurity: 98%