Learn More
Medchemexpress LLC ODN 1018 | 937402-51-2 | DNA, d(P-thio)(T-G-A-C-T-G-T-G-A-A-C-G-T-T-C-G-A-G-A-T-G-A) | 1 MG
SDP
Catalog No. 5000375926
Encompass_Preferred
ODN 1018 (1018 ISS) is a TLR9 agonist and immune modulator. It exhibits adjuvant activity and augments CD8+ T cell responses with LNP-encapsulated OVA peptides. It triggers sustained suppression of allergic airway inflammation and can be used for the research of allergic asthma and systemic lupus erythematosus.
  • TLR9 agonist and immune modulator
  • Exhibits adjuvant activity
  • Augments CD8+ T cell responses with LNP-encapsulated OVA peptides
  • Triggers sustained suppression of allergic airway inflammation
  • Used for research of allergic asthma
  • Used for research of systemic lupus erythematosus
Catalog No. 50-003-75926 Supplier Medchemexpress LLC Supplier No. HY1507241MG
ODN 1018 (1018 ISS) is a TLR9 agonist and immune modulator. It exhibits adjuvant activity and augments CD8+ T cell responses with LNP-encapsulated OVA peptides. It triggers sustained suppression of allergic airway inflammation and can be used for the research of allergic asthma and systemic lupus erythematosus.
  • TLR9 agonist and immune modulator
  • Exhibits adjuvant activity
  • Augments CD8+ T cell responses with LNP-encapsulated OVA peptides
  • Triggers sustained suppression of allergic airway inflammation
  • Used for research of allergic asthma
  • Used for research of systemic lupus erythematosus