Learn More
Medchemexpress LLC ODN 1018 sodium | 937402-51-2 | 1 MG
SDP
Catalog No. 5000375444
Encompass_Preferred
ODN 1018 sodium is an oligodeoxynucleotide and a TLR-9 agonist. It is a synthetic immunostimulatory sequence used as a vaccine adjuvant. It enhances cytotoxic effector mechanisms and stimulates anti-tumor immunity.
  • Enhances cytotoxic effector mechanisms
  • Stimulates anti-tumor immunity
  • Can be used for research of relapsed or refractory follicular lymphoma
  • Used as an adjuvant in the hepatitis B vaccine
  • Induces strong humoral and cellular immunity against HBV
  • Augments CD8+ T cell responses with LNP-encapsulated OVA peptides
  • Triggers sustained suppression of allergic airway inflammation
  • Can be used for research in allergic asthma and systemic lupus erythematosus
Catalog No. 50-003-75444 Supplier Medchemexpress LLC Supplier No. HY150724C1MG
ODN 1018 sodium is an oligodeoxynucleotide and a TLR-9 agonist. It is a synthetic immunostimulatory sequence used as a vaccine adjuvant. It enhances cytotoxic effector mechanisms and stimulates anti-tumor immunity.
  • Enhances cytotoxic effector mechanisms
  • Stimulates anti-tumor immunity
  • Can be used for research of relapsed or refractory follicular lymphoma
  • Used as an adjuvant in the hepatitis B vaccine
  • Induces strong humoral and cellular immunity against HBV
  • Augments CD8+ T cell responses with LNP-encapsulated OVA peptides
  • Triggers sustained suppression of allergic airway inflammation
  • Can be used for research in allergic asthma and systemic lupus erythematosus