Learn More
Medchemexpress LLC ODN 1585 | 386832-46-8 | DNA, d(G-sp-G-G-G-T-C-A-A-C-G-T-T-G-A-G-G-sp-G-sp-G-sp-G-sp-G) | 1 MG
SDP
Catalog No. 5000375458
Encompass_Preferred
ODN 1585 is a potent inducer of IFN and TNFα production and a strong stimulator of NK (natural killer) function. It increases CD8+ T-cell function, including the CD8+ T cell-mediated production of IFN-γ. ODN 1585 has been shown to induce regression of established melanomas in mice and can confer complete protection against malaria in mice. It is used for acute myelogenous leukemia (AML) and malaria research, and can also serve as a vaccine adjuvant.
  • Potent inducer of IFN and TNFα production
  • Potent stimulator of NK (natural killer) function
  • Increases CD8+ T-cell function, including IFN-γ production
  • Induces regression of established melanomas in mice
  • Confers complete protection against malaria in mice
  • Can be used for acute myelogenous leukemia (AML) and malaria research
  • Can be used as a vaccine adjuvant
Catalog No. 50-003-75458 Supplier Medchemexpress LLC Supplier No. HY1507251MG
ODN 1585 is a potent inducer of IFN and TNFα production and a strong stimulator of NK (natural killer) function. It increases CD8+ T-cell function, including the CD8+ T cell-mediated production of IFN-γ. ODN 1585 has been shown to induce regression of established melanomas in mice and can confer complete protection against malaria in mice. It is used for acute myelogenous leukemia (AML) and malaria research, and can also serve as a vaccine adjuvant.
  • Potent inducer of IFN and TNFα production
  • Potent stimulator of NK (natural killer) function
  • Increases CD8+ T-cell function, including IFN-γ production
  • Induces regression of established melanomas in mice
  • Confers complete protection against malaria in mice
  • Can be used for acute myelogenous leukemia (AML) and malaria research
  • Can be used as a vaccine adjuvant