Learn More
Medchemexpress LLC ODN 1585 (sodium) | DNA, d(G-sp-G-G-G-T-C-A-A-C-G-T-T-G-A-G-G-sp-G-sp-G-sp-G-sp-G), sodium salt | 1 MG
SDP
Catalog No. 5000374956
Encompass_Preferred
ODN 1585 (sodium) is a potent inducer of IFN and TNFα production, and a strong stimulator of NK (natural killer) cell function. It increases CD8+ T-cell function, including IFN-γ production. This product has been shown to induce regression of established melanomas and offer complete protection against malaria in mice. It can be used for acute myelogenous leukemia (AML) and malaria research, and also serves as a vaccine adjuvant.
  • Potent inducer of IFN and TNFα production
  • Strong stimulator of natural killer (NK) cell function
  • Increases CD8+ T-cell function and IFN-γ production
  • Induces regression of established melanomas
  • Confers protection against malaria
  • Applicable for acute myelogenous leukemia (AML) and malaria research
  • Functions as a vaccine adjuvant
  • Induces PBMC to produce IFN-α
  • Stimulates NK cell-mediated lysis of K562 cells
Catalog No. 50-003-74956 Supplier Medchemexpress LLC Supplier No. HY150725C1MG
ODN 1585 (sodium) is a potent inducer of IFN and TNFα production, and a strong stimulator of NK (natural killer) cell function. It increases CD8+ T-cell function, including IFN-γ production. This product has been shown to induce regression of established melanomas and offer complete protection against malaria in mice. It can be used for acute myelogenous leukemia (AML) and malaria research, and also serves as a vaccine adjuvant.
  • Potent inducer of IFN and TNFα production
  • Strong stimulator of natural killer (NK) cell function
  • Increases CD8+ T-cell function and IFN-γ production
  • Induces regression of established melanomas
  • Confers protection against malaria
  • Applicable for acute myelogenous leukemia (AML) and malaria research
  • Functions as a vaccine adjuvant
  • Induces PBMC to produce IFN-α
  • Stimulates NK cell-mediated lysis of K562 cells