Learn More
Enzo Life Sciences ODN 1668 (TLRGRADE™) (synthetic) (100µg)
SDP
Catalog No. 502009659
Encompass_Preferred
CpG oligodeoxynucleotide (Type B) with phosphorothioate backbone. Specific ligand for mouse TLR9 (Toll-like receptor 9). Formulation: Lyophilized. Sterile. Long Term Storage: +4°C.
Catalog No. 50-200-9659 Supplier Enzo Life Sciences Supplier No. ALX746001C100
CpG oligodeoxynucleotide (Type B) with phosphorothioate backbone. Specific ligand for mouse TLR9 (Toll-like receptor 9). Formulation: Lyophilized. Sterile. Long Term Storage: +4°C.