Learn More
Enzo Life Sciences ODN 1668 (TLRGRADE™) (synthetic) (BULK) (1mg)
SDP
Catalog No. 502009665
Encompass_Preferred
CpG oligodeoxynucleotide (Type B) with phosphorothioate backbone. Specific ligand for mouse TLR9 (Toll-like receptor 9). Formulation: Lyophilized. Sterile. Long Term Storage: +4°C.
Catalog No. 50-200-9665 Supplier Enzo Life Sciences Supplier No. ALX746051M001
CpG oligodeoxynucleotide (Type B) with phosphorothioate backbone. Specific ligand for mouse TLR9 (Toll-like receptor 9). Formulation: Lyophilized. Sterile. Long Term Storage: +4°C.