Learn More
Medchemexpress LLC Odn 1826 | 202668-42-6 | 10MG
SDP
Catalog No. 5000259918
Encompass_Preferred
ODN 1826 is a class B CpG oligodeoxynucleotide that functions as a mouse TLR9 agonist. It is a synthetic, single-stranded 20-mer with a full phosphorothioate backbone and unmethylated CpG motifs, used to activate B cells and induce pro-inflammatory cytokines in preclinical mouse studies.
  • Synthetic 20-mer oligodeoxynucleotide with a phosphorothioate backbone
  • Contains unmethylated CpG motifs that activate mouse TLR9
  • Potent stimulator of B cells and pro-inflammatory cytokine production
  • Commonly used as a vaccine adjuvant or immunostimulant in mouse models
  • Nuclease resistant for improved in vivo stability
  • High purity (>98% HPLC) available and lot-tested for biological activity
Catalog No. 50-002-59918 Supplier Medchemexpress LLC Supplier No. HY14624510MG
ODN 1826 is a class B CpG oligodeoxynucleotide that functions as a mouse TLR9 agonist. It is a synthetic, single-stranded 20-mer with a full phosphorothioate backbone and unmethylated CpG motifs, used to activate B cells and induce pro-inflammatory cytokines in preclinical mouse studies.
  • Synthetic 20-mer oligodeoxynucleotide with a phosphorothioate backbone
  • Contains unmethylated CpG motifs that activate mouse TLR9
  • Potent stimulator of B cells and pro-inflammatory cytokine production
  • Commonly used as a vaccine adjuvant or immunostimulant in mouse models
  • Nuclease resistant for improved in vivo stability
  • High purity (>98% HPLC) available and lot-tested for biological activity