Learn More
Invivogen ODN 1826 Biotin | 2x50ug
SDP
Catalog No. NC1367075
Encompass
ODN 1826 Biotin can be used to evaluate CpG ODN cellular uptake and localization using a biotin detection system and light microscopy.
Catalog No. NC1367075 Supplier Invivogen Supplier No. TLRL1826B
ODN 1826 Biotin can be used to evaluate CpG ODN cellular uptake and localization using a biotin detection system and light microscopy.