Learn More
Medchemexpress LLC Odn 1826 (sodium) | 202668-42-6 | 99.7% | 6364.1 | 5 MG
SDP
Catalog No. 5000221056
Encompass_Preferred
ODN 1826 (CpG 1826) is a class B CpG oligodeoxynucleotide that functions as a TLR9 agonist for research use. Supplied as a solid (sodium salt), it is used to stimulate innate immune responses, induce nitric oxide and iNOS production, and enhance apoptosis and immune surveillance in preclinical models.
  • Class B CpG oligodeoxynucleotide that activates TLR9.
  • Stimulates innate immune responses and enhances immune surveillance.
  • Induces nitric oxide (NO) and inducible nitric oxide synthase (iNOS) production.
  • Used in tumor immunology, vaccine adjuvant research, and atherosclerosis models.
  • Available as a solid sodium salt with high reported purity.
Catalog No. 50-002-21056 Supplier Medchemexpress LLC Supplier No. HY1462455MG
ODN 1826 (CpG 1826) is a class B CpG oligodeoxynucleotide that functions as a TLR9 agonist for research use. Supplied as a solid (sodium salt), it is used to stimulate innate immune responses, induce nitric oxide and iNOS production, and enhance apoptosis and immune surveillance in preclinical models.
  • Class B CpG oligodeoxynucleotide that activates TLR9.
  • Stimulates innate immune responses and enhances immune surveillance.
  • Induces nitric oxide (NO) and inducible nitric oxide synthase (iNOS) production.
  • Used in tumor immunology, vaccine adjuvant research, and atherosclerosis models.
  • Available as a solid sodium salt with high reported purity.