Learn More
Invivogen ODN 2006 1MG
SDP
Supplier:  Invivogen TLRL20061
Encompass
Class B CpG oligonucleotide - Human TLR9 ligand. Contains unmethylated CpG dinucleotides in particular sequence contexts (CpG motifs). Present at a 20-fold greater frequency in bacterial DNA compared to mammalian DNA.
Catalog No. NC9902842