Learn More
Invivogen ODN 2006 control (ODN 2137), 1 mg
SDP
Catalog No. NC9902844
Encompass
ODN 2006 Control (ODN 2137) contains GpC dinucleotides instead of CpGs and can be used as a negative control for ODN 2006 (type B, with a preference for human TLR9).
Catalog No. NC9902844 Supplier Invivogen Supplier No. TLRL2006C1
ODN 2006 Control (ODN 2137) contains GpC dinucleotides instead of CpGs and can be used as a negative control for ODN 2006 (type B, with a preference for human TLR9).