Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ODN 2007 sodium | 455348-63-7 | [DNA, d(P-thio)(T-C-G-T-C-G-T-T-G-T-C-G-T-T-T-T-G-T-C-G-T-T), sodium salt] | 1 MG
SDP

Supplier:  Medchemexpress LLC HY150734A1MG

Encompass_Preferred

ODN 2007 sodium is a class B CpG oligodeoxynucleotide (ODN) and a Toll-like receptor (TLR) ligand. It functions as an immunomodulator and vaccine adjuvant, enhancing immune responses in mammals, fish, and humans. The sequence is 5'-TCGTCGTTGTCGTTTTGTCGTT-3'.

  • Purity of 99.69%
  • Appears as a solid
  • Color is white to off-white
  • Enhances immune responses
  • Functions as an immunomodulator
  • Acts as a vaccine adjuvant
  • TLR ligand
  • Induces chicken macrophage interferon IFN-γ and IFN-β expression in vitro
  • Increases phosphorylation of ERK2 and AKT, and stimulates NO production in vitro
  • Enhances immune response in zebrafish against Vibrio traumaticus infection in vivo
  • Reduces cumulative mortality rate in treated zebrafish group in vivo
  • Decreases expression levels of intestinal immune-related genes in vivo

Catalog No. 50-003-75661


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.