Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ODN 2088 sodium | DNA, d(P-thio)(T-C-C-T-G-G-C-G-G-G-G-A-A-G-T), sodium salt | 1 MG
SDP

Supplier:  Medchemexpress LLC HY150738C1MG

Encompass_Preferred

ODN 2088 sodium is a potent inhibitor of TLR3, TLR7, and TLR9, showing no cytotoxic effects and inhibiting the release of IFN-α and IL-6. This product is for research use only.

  • Purity: 98.82%
  • Molecular weight (free acid): 4878
  • Appearance: White to off-white solid
  • Soluble in H2O at ≥ 25 mg/mL

Catalog No. 50-003-74045


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.