Learn More
Medchemexpress LLC ODN 21158 (sodium) | DNA, d(P-thio)(C-C-T-G-G-C-G-G-G-G), sodium salt | 1 MG
SDP
Supplier:  Medchemexpress LLC HY150739A1MG
Encompass_Preferred
ODN 21158 sodium is a potent G-modified TLR3 and TLR9 inhibitor that effectively inhibits IFN-α secretion in a dose-dependent manner. It is supplied as a solid, white to off-white colored compound.
  • Potent G-modified TLR3 and TLR9 inhibitor
  • Inhibits IFN-α secretion
  • Supplied as a solid, white to off-white powder
  • Recommended storage for solid at -20°C, sealed and away from moisture. In solvent, store at -80°C for up to 6 months or -20°C for 1 month.
Catalog No. 50-003-75459