Learn More
Medchemexpress LLC ODN 2216 | 332437-00-0 | DNA, d(G-sp-G-sp-G-G-G-A-C-G-A-T-C-G-T-C-G-sp-G-sp-G-sp-G-sp-G-sp-G) | 1 MG
SDP
Catalog No. 5000375741
Encompass_Preferred
ODN 2216 is a human-specific TLR9 (toll-like receptor 9) ligand or agonist. It induces high amounts of IFN-α and IFN-β, and IFN-α by pDC (plasmacytoid DC) and IL-12 (p40) production by DC (dendritic cells). It stimulates IFN-γ production in peripheral blood mononuclear cells (PBMC), indirectly mediated by IFN-α/β.
  • Activates NK cells and promotes IFN-γ production of TCR-triggered CD4+ T cells.
  • Drives Th1 development of naive CD4 T cells.
  • Significantly decreases apoptosis of thymic DC.
  • Induces T-cell mediated antitumor immune response in murine vaccination models.
Catalog No. 50-003-75741 Supplier Medchemexpress LLC Supplier No. HY1507411MG
ODN 2216 is a human-specific TLR9 (toll-like receptor 9) ligand or agonist. It induces high amounts of IFN-α and IFN-β, and IFN-α by pDC (plasmacytoid DC) and IL-12 (p40) production by DC (dendritic cells). It stimulates IFN-γ production in peripheral blood mononuclear cells (PBMC), indirectly mediated by IFN-α/β.
  • Activates NK cells and promotes IFN-γ production of TCR-triggered CD4+ T cells.
  • Drives Th1 development of naive CD4 T cells.
  • Significantly decreases apoptosis of thymic DC.
  • Induces T-cell mediated antitumor immune response in murine vaccination models.