Learn More
Medchemexpress LLC ODN 2216 sodium | 98.6% | 6432 (free acid) | 10 MG
SDP
Catalog No. 5000287162
Encompass_Preferred
ODN 2216 sodium is a synthetic oligodeoxynucleotide (sodium salt) that functions as a human-specific TLR9 ligand. It strongly induces type I interferons (IFN-α and IFN-β) via plasmacytoid dendritic cells and promotes IL-12 and IFN-γ production in immune cell assays, making it useful for in vitro and ex vivo immunology research focused on innate immune activation.
  • Human-specific TLR9 agonist.
  • Induces high levels of IFN-α and IFN-β.
  • Stimulates IL-12 production by dendritic cells.
  • Activates NK cells and promotes IFN-γ production in CD4+ T cells.
  • Supplied as the sodium salt suitable for in vitro use.
  • High purity (98.6%) and provided as a 10 MG quantity.
  • Oligonucleotide sequence provided to support experimental design.
Catalog No. 50-002-87162 Supplier Medchemexpress LLC Supplier No. HY150741C10MG
ODN 2216 sodium is a synthetic oligodeoxynucleotide (sodium salt) that functions as a human-specific TLR9 ligand. It strongly induces type I interferons (IFN-α and IFN-β) via plasmacytoid dendritic cells and promotes IL-12 and IFN-γ production in immune cell assays, making it useful for in vitro and ex vivo immunology research focused on innate immune activation.
  • Human-specific TLR9 agonist.
  • Induces high levels of IFN-α and IFN-β.
  • Stimulates IL-12 production by dendritic cells.
  • Activates NK cells and promotes IFN-γ production in CD4+ T cells.
  • Supplied as the sodium salt suitable for in vitro use.
  • High purity (98.6%) and provided as a 10 MG quantity.
  • Oligonucleotide sequence provided to support experimental design.