Learn More
Enzo Life Sciences ODN 2216 (TLRGRADE™) (synthetic) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX746005C100
Encompass_Preferred
CpG oligodeoxynucleotide (Type A) with mixed phosphodiester/phosphorothioate backbone. Specific ligand for human TLR9 (Toll-like receptor 9). Formulation: Lyophilized. Sterile. Long Term Storage: +4°C.
Catalog No. 50-200-9662