Learn More
Medchemexpress LLC ODN 2395 (sodium) | 1254617-22-5 | 98.2% | 7519.00 Da | C212H272N76O139P22 | 10 MG
SDP
Catalog No. 5000287264
Encompass_Preferred
ODN 2395 (sodium) is a synthetic C-class CpG oligodeoxynucleotide that acts as a Toll-like receptor 9 (TLR9) agonist and is used as an immunostimulatory agent and vaccine adjuvant in research. The product is supplied as a high-purity solid suitable for laboratory applications.
Catalog No. 50-002-87264 Supplier Medchemexpress LLC Supplier No. HY150743C10MG
ODN 2395 (sodium) is a synthetic C-class CpG oligodeoxynucleotide that acts as a Toll-like receptor 9 (TLR9) agonist and is used as an immunostimulatory agent and vaccine adjuvant in research. The product is supplied as a high-purity solid suitable for laboratory applications.