Learn More
Medchemexpress LLC Odn 2395 sodium | DNA, d(P-thio)(T-C-G-T-C-G-T-T-T-T-C-G-G-C-G-C-G-C-G-C-C-G), sodium salt | 5 MG
SDP
Catalog No. 5000375855
Encompass_Preferred
ODN 2395 sodium is a C class oligodeoxynucleotide that functions as a vaccine adjuvant and a TLR9 agonist. Its sequence is 5'-tcgtcgttttcggcgc:gcgccg-3'. It is intended for research use only.
  • C class oligodeoxynucleotide
  • Vaccine adjuvant
  • TLR9 agonist
Catalog No. 50-003-75855 Supplier Medchemexpress LLC Supplier No. HY150743C5MG
ODN 2395 sodium is a C class oligodeoxynucleotide that functions as a vaccine adjuvant and a TLR9 agonist. Its sequence is 5'-tcgtcgttttcggcgc:gcgccg-3'. It is intended for research use only.
  • C class oligodeoxynucleotide
  • Vaccine adjuvant
  • TLR9 agonist