Learn More
Medchemexpress LLC ODN 24987 sodium | DNA, d(P-thio)(C-C-T-G-G-C-c7G-G-G-G), sodium salt | 1 MG
SDP
Catalog No. 5000374841
Encompass_Preferred
ODN 24987 sodium is a Guanine-modified inhibitory oligonucleotide (ODN), targeting TLR9. It can inhibit IL-6 and IFN-α release and can be used for researching immune disorders. The sequence is 5'-C-C-T-G-G-C-c7G-G-G-G-3'.
  • Guanine-modified inhibitory oligonucleotide
  • Targets toll-like receptor 9 (TLR9)
  • Inhibits IL-6 and IFN-α release
  • Suitable for immune disorder research
  • Sequence: 5'-C-C-T-G-G-C-c7G-G-G-G-3'
Catalog No. 50-003-74841 Supplier Medchemexpress LLC Supplier No. HY150745A1MG
ODN 24987 sodium is a Guanine-modified inhibitory oligonucleotide (ODN), targeting TLR9. It can inhibit IL-6 and IFN-α release and can be used for researching immune disorders. The sequence is 5'-C-C-T-G-G-C-c7G-G-G-G-3'.
  • Guanine-modified inhibitory oligonucleotide
  • Targets toll-like receptor 9 (TLR9)
  • Inhibits IL-6 and IFN-α release
  • Suitable for immune disorder research
  • Sequence: 5'-C-C-T-G-G-C-c7G-G-G-G-3'