Learn More
Medchemexpress LLC ODN 4084-F sodium | 1934318-44-1 | DNA, d(P-thio)(C-C-T-G-G-A-T-G-G-G-A-A), sodium salt | 1 MG
SDP
Catalog No. 5000376792
Encompass_Preferred
ODN 4084-F sodium is a class B ('broadly-active') inhibitory ODN. It acts as a potent inhibitor of TLR9-induced B cells and macrophages. This product is a white to off-white solid.
  • Class B ('broadly-active') inhibitory ODN
  • Potent inhibitor of TLR9-induced B cells and macrophages
  • Solid appearance
  • White to off-white color
Catalog No. 50-003-76792 Supplier Medchemexpress LLC Supplier No. HY153839A1MG
ODN 4084-F sodium is a class B ('broadly-active') inhibitory ODN. It acts as a potent inhibitor of TLR9-induced B cells and macrophages. This product is a white to off-white solid.
  • Class B ('broadly-active') inhibitory ODN
  • Potent inhibitor of TLR9-induced B cells and macrophages
  • Solid appearance
  • White to off-white color