Learn More
Medchemexpress LLC ODN D-SL01 sodium | DNA, d(P-thio)(T-C-G-C-G-A-C-G-T-T-C-G-C-C-C-G-A-C-G-T-T-C-G-G-T-A), sodium salt | 1 MG
SDP
Catalog No. 5000374959
Encompass_Preferred
ODN D-SL01 sodium is a class B CpG oligodeoxynucleotide and a TLR9 agonist. It exhibits strong immunostimulatory activity in various vertebrate species and possesses anticancer activity.
  • Class B CpG oligodeoxynucleotide
  • TLR9 agonist
  • Strong immunostimulatory activity in various vertebrate species
  • Possesses anticancer activity
  • Oligonucleotide for cancer immunotherapy research
Catalog No. 50-003-74959 Supplier Medchemexpress LLC Supplier No. HY150748A1MG
ODN D-SL01 sodium is a class B CpG oligodeoxynucleotide and a TLR9 agonist. It exhibits strong immunostimulatory activity in various vertebrate species and possesses anticancer activity.
  • Class B CpG oligodeoxynucleotide
  • TLR9 agonist
  • Strong immunostimulatory activity in various vertebrate species
  • Possesses anticancer activity
  • Oligonucleotide for cancer immunotherapy research