Learn More
Medchemexpress LLC Odn D-SL03 (sodium) | 1198621-85-0 | 98.6% | 9345 (free acid) | 10 MG
SDP
Catalog No. 5000287158
Encompass_Preferred
ODN D-SL03 sodium is a C-class CpG oligodeoxynucleotide TLR9 agonist supplied as a solid for research use. It stimulates peripheral blood mononuclear cells (PBMCs) to produce interferon-α, activates human B cells, NK cells, and mononuclear cells, and modulates antigen-presentation markers. The oligonucleotide sequence is 5'-tcgcgaacgttcgccgcgttcgaacgcgg-3'.
  • Stimulates PBMCs to produce high levels of IFN-α.
  • Activates human B cells, NK cells, and mononuclear cells.
  • Up-regulates CD80, CD86, and HLA-DR expression on immune cells.
  • Sequence-defined oligonucleotide: 5'-tcgcgaacgttcgccgcgttcgaacgcgg-3'.
  • High purity (98.6%) suitable for research applications.
  • Supplied as a solid with recommended storage at -20°C; in solvent, -80°C up to 6 months.
  • Available in small amounts appropriate for assays (e.g., 10 mg).
  • For research use only.
Catalog No. 50-002-87158 Supplier Medchemexpress LLC Supplier No. HY150749A10MG
ODN D-SL03 sodium is a C-class CpG oligodeoxynucleotide TLR9 agonist supplied as a solid for research use. It stimulates peripheral blood mononuclear cells (PBMCs) to produce interferon-α, activates human B cells, NK cells, and mononuclear cells, and modulates antigen-presentation markers. The oligonucleotide sequence is 5'-tcgcgaacgttcgccgcgttcgaacgcgg-3'.
  • Stimulates PBMCs to produce high levels of IFN-α.
  • Activates human B cells, NK cells, and mononuclear cells.
  • Up-regulates CD80, CD86, and HLA-DR expression on immune cells.
  • Sequence-defined oligonucleotide: 5'-tcgcgaacgttcgccgcgttcgaacgcgg-3'.
  • High purity (98.6%) suitable for research applications.
  • Supplied as a solid with recommended storage at -20°C; in solvent, -80°C up to 6 months.
  • Available in small amounts appropriate for assays (e.g., 10 mg).
  • For research use only.