Learn More
Medchemexpress LLC ODN INH 18 sodium | DNA, d(P-thio)(C-C-T-G-G-A-T-G-G-G-A-A-C-T-T-A-C-C-G-C-T-G-C-A), sodium salt | 1 MG
SDP
Catalog No. 5000376409
Encompass_Preferred
ODN INH-18 sodium is a linear 24-mer class B INH-ODN. It shows inhibitory potency for TLR9 ligand-induced IFN-α production and is for research use only.
  • Linear 24-mer class B INH-ODN
  • 5' INH-ODN 4084-F sequence followed by a random stretch of 12 nucleotides
  • Lacks ability to form significant secondary structures
Catalog No. 50-003-76409 Supplier Medchemexpress LLC Supplier No. HY153840A1MG
ODN INH-18 sodium is a linear 24-mer class B INH-ODN. It shows inhibitory potency for TLR9 ligand-induced IFN-α production and is for research use only.
  • Linear 24-mer class B INH-ODN
  • 5' INH-ODN 4084-F sequence followed by a random stretch of 12 nucleotides
  • Lacks ability to form significant secondary structures