Learn More
Medchemexpress LLC Odn M362 | 934655-87-5 | DNA, d(P-thio)(T-C-G-T-C-G-T-C-G-T-T-C-G-A-A-C-G-A-C-G-T-T-G-A-T) | 1 MG
SDP
Catalog No. 5000375680
Encompass_Preferred
ODN M362 is a class C oligodeoxynucleotide that acts as a TLR-9 agonist. It can be utilized as a vaccine adjuvant and is known to induce cancer cell apoptosis.
  • TLR-9 agonist
  • Can be used as a vaccine adjuvant
  • Induces cancer cell apoptosis
Catalog No. 50-003-75680 Supplier Medchemexpress LLC Supplier No. HY150750A1MG
ODN M362 is a class C oligodeoxynucleotide that acts as a TLR-9 agonist. It can be utilized as a vaccine adjuvant and is known to induce cancer cell apoptosis.
  • TLR-9 agonist
  • Can be used as a vaccine adjuvant
  • Induces cancer cell apoptosis