Learn More
Invivogen ODN M362 - TLR9 ligand, 1 mg
SDP
Catalog No. NC9817208
Encompass
CpG ODNs are synthetic oligonucleotides that contain unmethylated CpG dinucleotides in particular sequence contexts (CpG motifs). These CpG motifs are present at a 20-fold greater frequency in bacterial DNA compared to mammalian DNA. CpG ODNs are recognized by Toll-like receptor 9 (TLR9) leading to strong immunostimulatory effects.
Catalog No. NC9817208 Supplier Invivogen Supplier No. TLRLM3621
CpG ODNs are synthetic oligonucleotides that contain unmethylated CpG dinucleotides in particular sequence contexts (CpG motifs). These CpG motifs are present at a 20-fold greater frequency in bacterial DNA compared to mammalian DNA. CpG ODNs are recognized by Toll-like receptor 9 (TLR9) leading to strong immunostimulatory effects.