Learn More
Invivogen Negative control for ODN M362, 200 µg
SDP
Catalog No. NC1586575
Encompass
ODN M362 Control contains GpC dinucleotides instead of CpGs and can be used as a negative control for ODN M362 (type C specific for human / mouse TLR9).
Catalog No. NC1586575 Supplier Invivogen Supplier No. TLRLM362
ODN M362 Control contains GpC dinucleotides instead of CpGs and can be used as a negative control for ODN M362 (type C specific for human / mouse TLR9).