Learn More
Invivogen ODN TTAGGG (A151), 1 mg
SDP
Supplier:  Invivogen TLRLTTAG1511
Encompass
ODN TTAGGG (A151), also known simply as A151, is a synthetic oligonucleotide (ODN) containing 4 repeats of the immunosuppressive TTAGGG motif commonly found in mammalian telomeric DNA. This ODN is also described as a cGAS inhibitor, acting through competition with DNA.
Catalog No. NC1310974