Learn More
Invivogen ODN2006-G5
SDP
Catalog No. NC9395503
Encompass
ODN2006-G5; 200ug; Stimulatory CpG ODN; Human TLR9 Ligand not stimulated by control; Endotoxin-Free
Catalog No. NC9395503 Supplier Invivogen Supplier No. TLRL2006G5
ODN2006-G5; 200ug; Stimulatory CpG ODN; Human TLR9 Ligand not stimulated by control; Endotoxin-Free