Learn More
Invivogen ODN 2006 (ODN 7909), 5 mg
SDP
Catalog No. NC9022164
Encompass
CpG ODNs are synthetic oligonucleotides that contain unmethylated CpG dinucleotides in particular sequence contexts (CpG motifs). These CpG motifs are present at a 20-fold greater frequency in bacterial DNA compared to mammalian DNA. CpG ODNs are recognized by Toll-like receptor 9 (TLR9) leading to strong immunostimulatory effects.
Catalog No. NC9022164 Supplier Invivogen Supplier No. TLRL20065
CpG ODNs are synthetic oligonucleotides that contain unmethylated CpG dinucleotides in particular sequence contexts (CpG motifs). These CpG motifs are present at a 20-fold greater frequency in bacterial DNA compared to mammalian DNA. CpG ODNs are recognized by Toll-like receptor 9 (TLR9) leading to strong immunostimulatory effects.