Learn More
Research Products International Corp Odo-Clave Deodorant Pads, Lemon Scent, 100 per Package
SDP
Catalog No. 50999240
Encompass
Eliminate foul autoclave odors with these unique pads. Patented technology pads contain a special heat released deodorant that neutralizes unpleasant odors generated during autoclaving. Easy to use, simply unwrap and place one pad in bag prior to autoclaving. Packaged 100 pads per case.
Catalog No. 50-999-240 Supplier Research Products International Corp Supplier No. 246140
Eliminate foul autoclave odors with these unique pads. Patented technology pads contain a special heat released deodorant that neutralizes unpleasant odors generated during autoclaving. Easy to use, simply unwrap and place one pad in bag prior to autoclaving. Packaged 100 pads per case.