Learn More
Research Products International Corp Odo-Clave Deodorant Pads, Pine Scent, 100 per Package
SDP
Catalog No. 50999239
Encompass
Eliminate foul autoclave odors with these unique pads. Patented technology pads contain a special heat released deodorant that neutralizes unpleasant odors generated during autclaving. Easy to use, simply unwarp and place one pad in bag prior to autoclaving. Packaged 100 pads per case.
Catalog No. 50-999-239 Supplier Research Products International Corp Supplier No. 246139
Eliminate foul autoclave odors with these unique pads. Patented technology pads contain a special heat released deodorant that neutralizes unpleasant odors generated during autclaving. Easy to use, simply unwarp and place one pad in bag prior to autoclaving. Packaged 100 pads per case.