Learn More
Medchemexpress LLC Odoroside A | 12738-19-1 | 1 MG
SDP
Catalog No. 5000417357
Encompass_Preferred
Odoroside A is an active ingredient extracted from the leaves of Nerium oleander Linn. It exhibits anti-cancer activity by inducing apoptosis and cell cycle arrest through the ROS/p53 signaling pathway, which leads to tumor cell death.
  • Extracted from Nerium oleander Linn
  • Exhibits anti-cancer activity
  • Induces apoptosis and cell cycle arrest
  • Acts through the ROS/p53 signaling pathway
  • Leads to tumor cell death
  • Purity of 99.73%
Catalog No. 50-004-17357 Supplier Medchemexpress LLC Supplier No. HYN74961MG
Odoroside A is an active ingredient extracted from the leaves of Nerium oleander Linn. It exhibits anti-cancer activity by inducing apoptosis and cell cycle arrest through the ROS/p53 signaling pathway, which leads to tumor cell death.
  • Extracted from Nerium oleander Linn
  • Exhibits anti-cancer activity
  • Induces apoptosis and cell cycle arrest
  • Acts through the ROS/p53 signaling pathway
  • Leads to tumor cell death
  • Purity of 99.73%