Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Odoroside A | 12738-19-1 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN749610MG

Encompass_Preferred

Odoroside A is an active ingredient extracted from the leaves of Nerium oleander Linn. It exhibits anti-cancer activity by inducing apoptosis and cell cycle arrest through the ROS/p53 signaling pathway, leading to tumor cell death.

  • Active ingredient extracted from Nerium oleander Linn
  • Exhibits anti-cancer activity
  • Induces apoptosis and cell cycle arrest
  • Works via the ROS/p53 signaling pathway
  • Leads to tumor cell death
  • High purity of 99.73%
  • Molecular weight: 518.68
  • Molecular formula: C₃₀H₄₆O₇
  • Appearance: White to off-white solid
  • Soluble in DMSO
  • Targeted pathway: Apoptosis
  • For research use only

Catalog No. 50-004-19101


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.