Learn More
StatLab OdorVault Formalin Strip
SDP
Catalog No. 22021382
Change view
Click to view available options
Type:
Formalin Absorbent
Catalog No. Type
22-021-382 Formalin Absorbent
1 options
Catalog No. 22-021-382 Supplier StatLab Supplier No. CPO1101P
Add to Cart
Add to Cart
OdorVault Formalin Strip is a revolutionary new strip that absorbs the worst odors
  • This strip can be placed onto or near any surfaces such as trashcans, bio-hazard bins, fume hoods and refrigerators
  • The OdorVault can be used in two ways - as an adhesive strip or as a box
  • Just remove the OdorVault from its package, peel the adhesive strip, and apply to any surface; or pierce the holes from the supplied box and place the strip and place wherever it s needed
  • Each strip will remove odors for up to 3 months
Specifications
Type Formalin Absorbent