Learn More
StatLab OdorVault Formalin Trap
Description
- Can be placed onto or near any surface, such as trashcans, bio-hazard bins, fume hoods, refrigerators, etc.
- Used as an adhesive strip or as a box.
- Each strip will remove odors for up to 3 months.
- Pack of 2 Units
Specifications
Specifications
| Product Type | Activated Carbon Solvent Trap Insert |
| Filter Type | Formaldehyde |
| Quantity | 2/Pk. |