Learn More
Medchemexpress LLC Odq | 41443-28-1 | MFCD00792620 | 99.8% | 187.16 g/mol | C9H5N3O2 | 10 MG
SDP
Catalog No. 5000212469
Encompass_Preferred
ODQ is a selective inhibitor of soluble guanylyl cyclase (sGC) used in biochemical and cell biology research to block nitric oxide-stimulated cyclic GMP production and to probe NO-cGMP signaling. It has been used to investigate sGC-dependent pathways and may enhance cisplatin-induced apoptosis in some cellular models.
  • Selective inhibitor of soluble guanylyl cyclase for NO-cGMP pathway studies.
  • High purity suitable for biochemical and cell-based assays.
  • Available as a powder and as pre-made DMSO solution for convenience.
  • Recommended storage supports multi-year stability for powder and long-term storage in solvent.
  • Supplied with SDS, COA, H NMR, and LC-MS data for quality control.
Catalog No. 50-002-12469 Supplier Medchemexpress LLC Supplier No. HY10125510MG
ODQ is a selective inhibitor of soluble guanylyl cyclase (sGC) used in biochemical and cell biology research to block nitric oxide-stimulated cyclic GMP production and to probe NO-cGMP signaling. It has been used to investigate sGC-dependent pathways and may enhance cisplatin-induced apoptosis in some cellular models.
  • Selective inhibitor of soluble guanylyl cyclase for NO-cGMP pathway studies.
  • High purity suitable for biochemical and cell-based assays.
  • Available as a powder and as pre-made DMSO solution for convenience.
  • Recommended storage supports multi-year stability for powder and long-term storage in solvent.
  • Supplied with SDS, COA, H NMR, and LC-MS data for quality control.