Learn More
AdipoGen ODQ (50 mg)
SDP
Supplier:  AdipoGen AGCR13500M050
Encompass_Preferred
Chemical. CAS: 41443-28-1. Formula: C9H5N3O2. MW: 187.2. Potent and highly selective, irreversible inhibitor of soluble guanylyl cyclase (sGC). The binding is competitive with nitric oxide (NO). Tool to elucidate the nitric oxide (NO)-cGMP pathway. Apoptosis inhibitor Inhibits growth and migration of prostate cancer cells independent of its effects on GMP levels.
Catalog No. 50-205-3875