Learn More
Abcam ODQ, NO-sensitive guanylyl cyclase inhibitor, 50MG
Catalog No. 502707926
Encompass_Preferred
MW 187.15 Da, Purity >99%. Selective, potent inhibitor of nitric oxide-sensitive guanylyl cyclase. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7926 Supplier Abcam Supplier No. AB12002250MG
MW 187.15 Da, Purity >99%. Selective, potent inhibitor of nitric oxide-sensitive guanylyl cyclase. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement