Learn More
WELCH MATERIALS INC Ultisil ODS-3, 5µm, 100Å, 4.6×250mm
Catalog No. NC2052214
Encompass_Preferred
Analytical Column, Ultisil ODS-3, 5μm, 100Å, 4.6×250mm, pH range: 2.0-8.0, Pore Size: 100Å, USP listing: L1, Carbon Loading: 15%, Surface Area: 380m2/g, endcapped.
Catalog No. NC2052214 Supplier WELCH MATERIALS INC Supplier No. H0027531043
Analytical Column, Ultisil ODS-3, 5μm, 100Å, 4.6×250mm, pH range: 2.0-8.0, Pore Size: 100Å, USP listing: L1, Carbon Loading: 15%, Surface Area: 380m2/g, endcapped.