Learn More
YMC America, Inc ODS-A 5UM 200A 100X4.6MM
SDP
Catalog No. 50157156
Encompass_Preferred
AA20S05-1046WT: ODS-A, 5um, 200A, 100X4.6mm Column
Catalog No. 50-157-156 Supplier YMC America, Inc Supplier No. AA20S051046WT
AA20S05-1046WT: ODS-A, 5um, 200A, 100X4.6mm Column