Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc ODS-A 5UM 300A 250X6MM
SDP

Catalog No. 5090004527
Encompass_Preferred

Ods-A 5Um; 300A 250 X 6mm

Catalog No. 50-900-04527 Supplier YMC America, Inc Supplier No. AA30S052506WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ods-A 5Um; 300A 250 X 6mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.